  • Search Results
  • Lengthening of high-yield production levels of monoclonal antibody-producing Chinese hamster ovary cells by downregulation of breast cancer 1.
Lengthening of high-yield production levels of monoclonal antibody-producing Chinese hamster ovary cells by downregulation of breast cancer 1.
Journal of bioscience and bioengineering (2016-10-16)
Rima Matsuyama, Noriko Yamano, Namiko Kawamura, Takeshi Omasa
ABSTRACT
The establishment process of high-producing Chinese hamster ovary (CHO) cells for therapeutic protein production is usually laborious and time consuming because of the low probability of obtaining stable, high-producing clones over a long term. Thus, development of an efficient approach is required to establish stable, high-producing cells. This study presents a novel method that can efficiently establish sustainably high-producing cell lines by acceleration of transgene amplification and suppression of transgene silencing. The effects of breast cancer 1 (BRCA1) downregulation on gene amplification efficiency and long-term productivity were investigated in CHO cells. Small interfering RNA expression vectors against BRCA1 were transfected into the CHO DG44-derived antibody-producing cell clone. Individual cell clones were obtained after induction of gene amplification in the presence of 400 nM methotrexate, which were cultured until passage 20. BRCA1-downregulated cell clones CHO B1Sa and B1Sb displayed 2.2- and 1.6-fold higher specific production rates than the S-Mock clone. Fluorescence in situ hybridization showed that transgene amplification occurred at a high frequency in B1Sa and B1Sb clones. Moreover, B1Sa and B1Sb clones at 20 passages had approximately 3.5- and 5.3-fold higher productivity than the S-Mock clone. Histone modification analysis revealed a decrease in an active mark for transcription, trimethylation of histone H3 at lysine 4 (H3K4), in the transgene locus of the S-Mock clone. However, H3K4 trimethylation levels were not decreased in B1Sa and B1Sb clones during long term culture. Our results suggest that high-producing cells, which maintain their productivity long-term, were efficiently established by BRCA1 downregulation.
Sigma-Aldrich
Lys-Lys-Lys-Lys, ≥95% (TLC)