  • Search Results
  • Differential regulation of E-cadherin and alpha-smooth muscle actin by BMP 7 in human renal proximal tubule epithelial cells and its implication in renal fibrosis.
Differential regulation of E-cadherin and alpha-smooth muscle actin by BMP 7 in human renal proximal tubule epithelial cells and its implication in renal fibrosis.
American journal of physiology. Renal physiology (2009-09-11)
Mangalakumar Veerasamy, Tri Q Nguyen, Reza Motazed, Alexander L Pearson, Roel Goldschmeding, Mark E C Dockrell
ABSTRACT
Chronic kidney diseases are characterized by progressive tubulointerstitial fibrosis, and TGFbeta1 plays a crucial role in its development. Bone morphogenic protein 7 (BMP 7), another member of the TGF superfamily, antagonized the profibrotic effects of TGFbeta1, including epithelial mesenchymal transition and E-cadherin loss, in the previous studies from animal models. We investigated the effect of BMP 7 on TGFbeta1-mediated E-cadherin loss in two different transformed human adult proximal tubule epithelia. We found that BMP 7 not only failed to prevent TGFbeta1-mediated E-cadherin loss but itself downregulated E-cadherin levels and that it had an additive effect with TGFbeta1 in inducing E-cadherin loss. The downregulation of E-cadherin by BMP 7 was mediated through the Smad1/5 pathway. BMP 7-mediated E-cadherin loss was not followed by de novo alpha-smooth muscle actin (alpha-SMA) expression (a marker of myofibroblastic phenotype), which was due to the concurrent induction of Inhibitor of DNA binding 1 (Id1, a basic helix loop helix class transcriptional regulator) through a non-Smad pathway. Concurrent treatment of BMP 7 and TGFbeta1 prevented TGFbeta1-mediated alpha-SMA induction. In summary, our results suggest that E-cadherin loss, the key feature of epithelial mesenchymal transition, will not necessarily be followed by total phenotype change; rather, cells may undergo some loss of phenotypic marker in a ligand-dependent manner and participate in reparative processes. The inhibition of de novo expression of alpha-SMA could explain the antifibrotic effect of BMP 7. Id1 might play a crucial role in maintaining proximal tubule epithelial cell phenotype and its signaling regulation could be a potential therapeutic target.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Transforming Growth Factor-β1 human, TGF-β1, recombinant, expressed in CHO cells, powder, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution