  • Search Results
  • The 22Rv1 prostate cancer cell line carries mixed genetic ancestry: Implications for prostate cancer health disparities research using pre-clinical models.
The 22Rv1 prostate cancer cell line carries mixed genetic ancestry: Implications for prostate cancer health disparities research using pre-clinical models.
The Prostate (2017-10-17)
Leanne Woods-Burnham, Anamika Basu, Christina K Cajigas-Du Ross, Arthur Love, Clayton Yates, Marino De Leon, Sourav Roy, Carlos A Casiano
ABSTRACT
Understanding how biological factors contribute to prostate cancer (PCa) health disparities requires mechanistic functional analysis of specific genes or pathways in pre-clinical cellular and animal models of this malignancy. The 22Rv1 human prostatic carcinoma cell line was originally derived from the parental CWR22R cell line. Although 22Rv1 has been well characterized and used in numerous mechanistic studies, no racial identifier has ever been disclosed for this cell line. In accordance with the need for racial diversity in cancer biospecimens and recent guidelines by the NIH on authentication of key biological resources, we sought to determine the ancestry of 22RV1 and authenticate previously reported racial identifications for four other PCa cell lines. We used 29 established Ancestry Informative Marker (AIM) single nucleotide polymorphisms (SNPs) to conduct DNA ancestry analysis and assign ancestral proportions to a panel of five PCa cell lines that included 22Rv1, PC3, DU145, MDA-PCa-2b, and RC-77T/E. We found that 22Rv1 carries mixed genetic ancestry. The main ancestry proportions for this cell line were 0.41 West African (AFR) and 0.42 European (EUR). In addition, we verified the previously reported racial identifications for PC3 (0.73 EUR), DU145 (0.63 EUR), MDA-PCa-2b (0.73 AFR), and RC-77T/E (0.74 AFR) cell lines. Considering the mortality disparities associated with PCa, which disproportionately affect African American men, there remains a burden on the scientific community to diversify the availability of biospecimens, including cell lines, for mechanistic studies on potential biological mediators of these disparities. This study is beneficial by identifying another PCa cell line that carries substantial AFR ancestry. This finding may also open the door to new perspectives on previously published studies using this cell line.
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Insulin from bovine pancreas, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture