Merck
HomeTitration & Karl Fischer AnalysisDetermination of Water Content in Ethyl acetate, acetic acid ethyl ester Using Karl Fischer Titration
Determination of Water Content By Karl Fischer Method in Ethyl Acetate
General Information for Water Determination in Esters of Carboxylic Acids
Esters of carboxylic acid can be analyzed analogously to the hydrocarbons according to the standard Karl Fischer method. Long-chain compounds require the addition of solubilisers, e.g., long-chain alcohols or chloroform. Unsaturated compounds can usually be titrated in the same way. Recommended methods are both the Karl Fischer volumetric titration with one or two component reagents, as well as the coulometric analysis. The latter is predominantly applied for low water concentrations (< 0.1%).
Methods for Water Determination in Ethyl Acetate sample
Ethyl acetate is easily soluble in alcoholic media. Water determination can be carried out without problems according to standard methods.
One-Component Volumetric Karl Fischer Titration
Karl Fischer Reagents:
  • Karl Fischer Titrant
    Aquastar® - CombiTitrant 5 (1.88005) - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 5 mg water
    or
    Aquastar® - CombiTitrant 2 (1.88002) - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 2 mg water
  • Solvent
    50 mL Aquastar® - CombiSolvent (1.88008) - Methanol-free solvent for volumetric Karl Fischer titration with one component reagents
    or
    50 mL Aquastar® - CombiMethanol (1.88009) - Solvent for volumetric Karl Fischer titration with one component reagents, max. 0.01 % water
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
10 mL
Titration Procedure:
The titration medium is first placed into the titration cell and titrated dry by means of the titrant. Then the sample is added with a syringe (exact sample weight determination by weighing the syringe before and after injection) or volumetric pipette, and the titration is started.
Two-Component Volumetric Karl Fischer Titration
Karl Fischer Reagents:
  • Karl Fischer Titrant
    Aquastar® - Titrant 5 (1.88010) - Titrant for volumetric titration with two component reagents, 1 mL = approx. 5 mg water
    or
    Aquastar® - Titrant 2 (1.88011) - Titrant for volumetric titration with two component reagents, 1 mL = approx. 2 mg water
  • Karl Fischer Solvent
    null Aquastar® - Solvent (1.88015) - Solvent for volumetric titration with two component reagents
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
10 mL
Titration Procedure:
The titration medium is first placed into the titration cell and titrated dry by means of the titrant. Then the sample is added with a syringe (exact sample weight determination by weighing the syringe before and after injection) or volumetric pipette, and the titration is started.
Coulometric Karl Fischer Titration (With Diaphragm)
Karl Fischer Reagents:
  • Catholyte
    5 mL Aquastar® - CombiCoulomat frit (1.09255) - Coulometric Karl Fischer reagent for cells with diaphragm
  • Anolyte
    100 mL Aquastar® - CombiCoulomat frit (1.09255) - Coulometric Karl Fischer reagent for cells with diaphragm
Titration Parameters:
Default coulometer settings for cells with a diaphragm:
For end point indication, e.g.:
I(pol) = 5 - 10 µA, U(EP) = 50 - 100 mV
Stop criterion: drift < 10 µg/min
Sample size:
1 mL
Titration Procedure:
The Karl-Fischer reagent is placed into the cathode and anode compartments of the titration cell with a diaphragm. The coulometer is started, and the solvent is titrated dry. After preliminary titration and stabilization of drift, the sample is injected into the titration cell with a syringe (exact sample weight determination by weighing the syringe before and after injection), and the water determination is started.
Coulometric Karl Fischer Titration (Without Diaphragm)
Karl Fischer Reagents:
  • Working medium
    100 mL Aquastar® - CombiCoulomat fritless (1.09257) - Coulometric Karl Fischer reagent for cells with or without diaphragm
Titration Parameters:
Default coulometer settings for cells without a diaphragm:
For end point indication, e.g.:
I(pol) = 5 - 10 µA, U(EP) = 50 - 100 mV
Stop criterion: drift < 10 µg/min
Sample size:
1 mL
Titration Procedure:
The Karl-Fischer reagent is placed into the titration cell without a diaphragm. The coulometer is started, and the solvent is titrated dry. After preliminary titration and stabilization of drift, the sample is injected into the titration cell with a syringe (exact sample weight determination by weighing of syringe before and after injection), and the water determination is started.