Merck
HomeSoil, Solid Waste & Groundwater TestingAnalytical Method: Nickel (Total) in Electrophoretic Paints
Analytical Method: Nickel (Total) in Electrophoretic Paints
Instruments and Reagents
  • Product No. 173016 Spectroquant® VIS Spectrophotometer Prove 100 or
  • Product No. 173017 Spectroquant® UV/VIS Spectrophotometer Prove 300 or
  • Product No. 173018 Spectroquant® UV/VIS Spectrophotometer Prove 600 or
  • Product No. 109748 Spectroquant® Photometer NOVA 30 or
  • Product No. 109751 Spectroquant® Photometer NOVA 60 or
  • Product No. 173632 Spectroquant® Colorimeter Move 100
  • Product No. 114946 Rectangular cells 10 mm and/or
  • Product No. 114947 Rectangular cells 20 mm and/or
  • Product No. 114944 Rectangular cells 50 mm
  • Product No. 114554 Spectroquant® Nickel Cell Test
  • Product No. 114785 Spectroquant® Nickel Test
  • Product No. 100731 Sulfuric acid 95 - 97 % for analysis
  • Product No. 107210 Hydrogen peroxide 30 % (Perhydrol®) for analysis
  • Product No. 105590 Sodium hydroxide solution 32 %
  • Product No. 109535 MColorpHastTM Universal indicator strips pH 0 - 14
  • Product No. 116754 Water for analysis
Sample Preparation
In a 400-mL glass beaker add 10 mL homogenized paint sample to 10 mL of sulfuric acid 95 - 97 % for analysis, swirling to mix. For complete solubilisation of the sedimented resins, heat the mixture on the steambath to
100 °C. After addition of 2 mL Perhydrol®, the dark-brown solution is decolorized. When foaming has subsided, add once more 2 mL of Perhydrol®, followed by a slow and cautious warming. After this add further 2 mL Perhydrol® and heat to boiling (glass beads, as anti-bump medium).
If the mixture remains clear and colorless after 10 minutes heating, allow to cool to room temperature. Should the color remain brown, repeat the treatment with 2 mL Perhydrol® with a further 10 minutes heating.
Dilute the cooled solution to 100 mL with water for analysis and adjust the pH (pH-meter) to 4, using sodium hydroxide solution 32 % and transfer to a 200-mL standard volumetric flask. Fill up to volume with water for analysis and mix well.
Analysis
Determine with the above mentioned test kits.
Calculation
Using the 1+99 sample dilution the analysis value corresponds directly to the nickel (total) content in mg/l.