Merck
HomeOrganic ElectronicsFabrication of Poly(triaryl amine) Field-effect Transistors
Fabrication of Poly(triaryl amine) Field-effect Transistors
PTAA Description
Poly[bis(4-phenyl)(2,4,6-trimethylphenyl)amine] (PTAA; 702471) is an amorphous p-type polymer semiconductor.1, 2 It may be used to fabricate field-effect transistors (FETs). PTAA is also useful as a hole transport material in organic light emitting diodes (OLEDs).
Poly[bis(4-phenyl)(2,4,6-trimethylphenyl)amine]
Procedure for Fabrication of PTAA Field-effect Transistors
Bottom-gate bottom-contact field-effect transistors (FETs) were fabricated in a nitrogen atmosphere on highly doped Si-wafers with a thermally grown 250 nm SiO2 layer. The two layers served as the gate electrode and gate insulator. Au source and drain electrodes (30 nm thick) were defined by standard photolithography:
channel length (L) = 10 mm
channel width (W) = 10 mm
Before deposition of the PTAA polymer layer, the Si-wafers were treated with octyltrichlorosilane (OTS-18; 104817 ) by immersing them in 10 mM solutions in toluene for 15 minutes at 60 °C.
A homogeneous solution of PTAA was prepared in toluene at room temperature containing 1.0 wt% of the polymer. This solution was deposited via spin-coating at 500 rpm for 30 seconds followed by 2,000 rpm for 50 seconds.
Results
Electrical characterization of the PTAA FETs was conducted in a nitrogen atmosphere with a HP4155B semiconductor parameter analyzer. Field-effect mobilities were calculated from transfer characteristics (saturation regime) employing the relation:3
Equation
Isd is the source-drain current (saturation regime)
Vg and Vsd gate and soxce-drain voltage, respectively
Ci the insulator capacitance
W and L the channel width and length
V0 the turn-on voltage
Transfer and output curves for PTAA transistors are shown in Figures 1 and 2.
Transfer output curves for PTAA transistors corresponding to field effect mobility of 4 x 10–3 cm2/Vs.
Figure 1.Transfer output curves for PTAA transistors corresponding to field effect mobility of 4 x 10–3 cm2/Vs.
Output curves for PTAA transistors corresponding to field effect mobility of 4 x 10–3 cm2/Vs.
Figure 2.Output curves for PTAA transistors corresponding to field effect mobility of 4 x 10–3 cm2/Vs.
Materials
Loading
References
1.
Chabinyc ML, Jimison LH, Rivnay J, Salleo A. 2008. Connecting Electrical and Molecular Properties of Semiconducting Polymers for Thin-Film Transistors. MRS Bull.. 33(7):683-689. http://dx.doi.org/10.1557/mrs2008.140
2.
Veres J, Ogier S, Leeming S, Cupertino D, Mohialdin Khaffaf S. 2003. Low-k Insulators as the Choice of Dielectrics in Organic Field-Effect Transistors. Adv. Funct. Mater.. 13(3):199-204. http://dx.doi.org/10.1002/adfm.200390030
3.
Brown A, Jarrett C, de Leeuw D, Matters M. 1997. Field-effect transistors made from solution-processed organic semiconductors. Synthetic Metals. 88(1):37-55. http://dx.doi.org/10.1016/s0379-6779(97)80881-8
Data courtesy of Dr. Iain McCulloch, Imperial College London and Flexink, Inc.