Merck
HomeNext-Generation SequencingPeptide Modifications: N-Terminal, Internal, and C-Terminal
Peptide Modifications: N-Terminal, Internal, and C-Terminal
1.0 N-Terminal Modifications
1.1  Acetylation
This modification removes the positive charge on the N-terminal of peptides, thus mimicking natural proteins. In some cases, it increases peptide stability by preventing N-terminal degradation 1, 2
Acetylation
References
1.
Arnesen T. Towards a Functional Understanding of Protein N-Terminal Acetylation. PLoS Biol. 9(5):e1001074. http://dx.doi.org/10.1371/journal.pbio.1001074
2.
Wallace RJ. 1992. Acetylation of peptides inhibits their degradation by rumen micro-organisms. Br J Nutr. 68(2):365-372. http://dx.doi.org/10.1079/bjn19920095
1.2 Biotin
Biotin has a very strong affinity for streptavidin and avidin. Biotin-labeled peptides are commonly used in immunoassays1, histocytochemistry2, and fluorescence based flow cytometry3
Biotin
References
1.
Sélo I, Négroni L, Créminon C, Grassi J, Wal J. 1996. Preferential labeling of ?-amino N-terminal groups in peptides by biotin: application to the detection of specific anti-peptide antibodies by enzyme immunoassays. Journal of Immunological Methods. 199(2):127-138. http://dx.doi.org/10.1016/s0022-1759(96)00173-1
2.
HOWL J, WANG X, KIRK CJ, WHEATLEY M. 1993. Fluorescent and biotinylated linear peptides as selective bifunctional ligands for the V1a vasopressin receptor. Eur J Biochem. 213(2):711-719. http://dx.doi.org/10.1111/j.1432-1033.1993.tb17811.x
3.
Buranda, et. al.. 1991. Peptide, antibodies and FRET on beads in flow cytometry: a model system using fluoresceinated and biotinylated β-endorphin. Cytometry.(37):21-31. https://doi.org/10.1002/(SICI)1097-0320(19990901)37:1<21::AID-CYTO3>3.0.CO;2-G
1.3  Dansyl
Dansyl labeled peptides are used in fluorescence based assays.
Dansyl
References
1.
Glukhov E, Stark M, Burrows LL, Deber CM. 2005. Basis for Selectivity of Cationic Antimicrobial Peptides for BacterialVersusMammalian Membranes. J. Biol. Chem.. 280(40):33960-33967. http://dx.doi.org/10.1074/jbc.m507042200
2.
Matsuzaki K, Mitani Y, Akada K, Murase O, Yoneyama S, Zasloff M, Miyajima K. 1998. Mechanism of Synergism between Antimicrobial Peptides Magainin 2 and PGLa?. Biochemistry. 37(43):15144-15153. http://dx.doi.org/10.1021/bi9811617
3.
Pecht I, Maron E, Arnon R, Sela M. 1971. Specific Excitation Energy Transfer from Antibodies to Dansyl-Labeled Antigen. Studies with the "Loop" Peptide of Hen Egg-White Lysozyme. Eur J Biochem. 19(3):368-371. http://dx.doi.org/10.1111/j.1432-1033.1971.tb01325.x
1.4 2, 4-Dinitrophenyl
2, 4-DNP is used as a quencher for (7-methoxy coumarin-4-yl) acetyl (MCA) and sometimes tryptophan. This modification can be attached at the N-terminal of a peptide or as an internal modification through lysine side chain.
2, 4-Dinitrophenyl
References
1.
Vickers C, Hales P, Kaushik V, Dick L, Gavin J, Tang J, Godbout K, Parsons T, Baronas E, Hsieh F, et al. 2002. Hydrolysis of Biological Peptides by Human Angiotensin-converting Enzyme-related Carboxypeptidase. J. Biol. Chem.. 277(17):14838-14843. http://dx.doi.org/10.1074/jbc.m200581200
2.
Knight C, Willenbrock F, Murphy G. 1992. A novel coumarin-labelled peptide for sensitive continuous assays of the matrix metalloproteinases. 296(3):263-266. http://dx.doi.org/10.1016/0014-5793(92)80300-6
1.5 Fluorescein
Fluorescein-labeled peptides have many fluorescein based biomolecular applications including protein-protein interaction, flow cytometry and localization studies1-3.
Fluorescein
References
1.
Richard JP, Melikov K, Vives E, Ramos C, Verbeure B, Gait MJ, Chernomordik LV, Lebleu B. 2003. Cell-penetrating Peptides. J. Biol. Chem.. 278(1):585-590. http://dx.doi.org/10.1074/jbc.m209548200
2.
Farley RA, Tran CM, Carilli CT, Hawke D, Shively JE. 1984. The amino acid sequence of a fluorescein-labeled peptide from the active site of (Na,K)-ATPase. J Biol Chem .. 15(259):9532-5.
3.
Futaki S, Suzuki T, Ohashi W, Yagami T, Tanaka S, Ueda K, Sugiura Y. 2001. Arginine-rich Peptides. J. Biol. Chem.. 276(8):5836-5840. http://dx.doi.org/10.1074/jbc.m007540200
4.
Foerg C, Weller KM, Rechsteiner H, Nielsen HM, Fernández-Carneado J, Brunisholz R, Giralt E, Merkle HP. 2008. Metabolic Cleavage and Translocation Efficiency of Selected Cell Penetrating Peptides: A Comparative Study with Epithelial Cell Cultures. AAPS J. 10(2):349-359. http://dx.doi.org/10.1208/s12248-008-9029-4
1.6 7-methoxycoumarin acetic acid (Mca)
7-methoxy coumarin-labeled peptides have applications in protein-protein interaction and localization studies1-3.
7-methoxycoumarin acetic acid (Mca)
References
1.
Vidal P, Chaloin L, Méry J, Lamb N, Lautredou N, Bennes R, Heitz F. 1996. Solid-phase Synthesis and Cellular Localization of a C- and/or N-terminal Labelled Peptide. J. Peptide Sci.. 2(2):125-133. http://dx.doi.org/10.1002/psc.58
2.
Yandek LE, Pokorny A, Florén A, Knoelke K, Langel Ü, Almeida PF. 2007. Mechanism of the Cell-Penetrating Peptide Transportan 10 Permeation of Lipid Bilayers. Biophysical Journal. 92(7):2434-2444. http://dx.doi.org/10.1529/biophysj.106.100198
1.7 Palmitic Acid
Palmitic acid is 16-carbon fatty acid that is conjugated to peptides to increase their cell permeability and help binding of the peptides to cell membrane 1-2.
Palmitic Acid
References
1.
Avrahami D, Shai Y. 2004. A New Group of Antifungal and Antibacterial Lipopeptides Derived from Non-membrane Active Peptides Conjugated to Palmitic Acid. J. Biol. Chem.. 279(13):12277-12285. http://dx.doi.org/10.1074/jbc.m312260200
2.
Buss JE, Sefton BM. 1986. Direct identification of palmitic acid as the lipid attached to p21ras.. Mol. Cell. Biol.. 6(1):116-122. http://dx.doi.org/10.1128/mcb.6.1.116
2.0 Internal Modifications
2.1 Cyclization (Disulfide Bonds)
This process involves formation of disulfide bond between two cysteine residues. Peptides can be cyclized to stabilize the peptide conformation, increase bioactivity, and enzyme stability1-2.
References
1.
Góngora-Benítez M, Tulla-Puche J, Albericio F. 2014. Multifaceted Roles of Disulfide Bonds. Peptides as Therapeutics. Chem. Rev.. 114(2):901-926. http://dx.doi.org/10.1021/cr400031z
2.
Schmelz EA, Huffaker A, Carroll MJ, Alborn HT, Ali JG, Teal PE. 2012. An Amino Acid Substitution Inhibits Specialist Herbivore Production of an Antagonist Effector and Recovers Insect-Induced Plant Defenses. Plant Physiol.. 160(3):1468-1478. http://dx.doi.org/10.1104/pp.112.201061
2.2 Cysteine Carbamidomethylation (CAM)
Carbamidomethylation (CAM) is a deliberate post-translational modification introduced to cysteine residues by reacting with iodoacetamide. Peptides with this modification are mainly used in Peptide Mass Fingerprinting for identification and characterization of proteins1. In other assays, this process is used to block Cysteine from oxidation2.
Cysteine Carbamidomethylation (CAM)
References
1.
Wilkins MR, Appel RD, Williams KL, Hochstrasser DF. 2007. Proteome Research. Berlin, Heidelberg: Springer Berlin Heidelberg.
2.
Rombouts I, Lagrain B, Brunnbauer M, Delcour JA, Koehler P. 2013. Improved identification of wheat gluten proteins through alkylation of cysteine residues and peptide-based mass spectrometry. Sci Rep. 3(1): http://dx.doi.org/10.1038/srep02279
2.3 Isotope labeled Amino Acids
AQUA peptides are synthetic peptides with amino acids enriched in 18O, 13C, and/or 14N. They are similar to their native peptides in terms of chemical, physical properties and also their biological activities1. Main applications for these peptides are to study protein interactions, proteins, post translation modifications such as ubiquitination and phosphorylation2-5.
References
1.
Kettenbach AN, Rush J, Gerber SA. 2011. Absolute quantification of protein and post-translational modification abundance with stable isotope?labeled synthetic peptides. Nat Protoc. 6(2):175-186. http://dx.doi.org/10.1038/nprot.2010.196
2.
Li H, Xu C, Blais S, Wan Q, Zhang H, Landry SJ, Hioe CE. 2009. Proximal Glycans Outside of the Epitopes Regulate the Presentation of HIV-1 Envelope gp120 Helper Epitopes. J Immunol. 182(10):6369-6378. http://dx.doi.org/10.4049/jimmunol.0804287
3.
Le Bihan T, Grima R, Martin S, Forster T, Le Bihan Y. 2010. Quantitative analysis of low-abundance peptides in HeLa cell cytoplasm by targeted liquid chromatography/mass spectrometry and stable isotope dilution: emphasising the distinction between peptide detection and peptide identification. Rapid Commun. Mass Spectrom.. 24(7):1093-1104. http://dx.doi.org/10.1002/rcm.4487
4.
Santhoshkumar P, Raju M, Sharma KK. ?A-Crystallin Peptide 66SDRDKFVIFLDVKHF80 Accumulating in Aging Lens Impairs the Function of ?-Crystallin and Induces Lens Protein Aggregation. PLoS ONE. 6(4):e19291. http://dx.doi.org/10.1371/journal.pone.0019291
5.
Sato Y, Miyashita A, Iwatsubo T, Usui T. 2012. Simultaneous Absolute Protein Quantification of Carboxylesterases 1 and 2 in Human Liver Tissue Fractions using Liquid Chromatography-Tandem Mass Spectrometry. Drug Metab Dispos. 40(7):1389-1396. http://dx.doi.org/10.1124/dmd.112.045054
6.
Brun V, Masselon C, Garin J, Dupuis A. 2009. Isotope dilution strategies for absolute quantitative proteomics. Journal of Proteomics. 72(5):740-749. http://dx.doi.org/10.1016/j.jprot.2009.03.007
2.4 Phosphorylation
Phosphorylation can be performed on Tyr, Ser and Thr residues as a posttranslational modification (PTM) on peptides. Phosphorylated peptides have application in many cellular processes such as gene expression, protein-protein interaction and signal transduction in plants and animals1,2.
Phosphorylation
References
1.
Hunter T. 2000. Signaling?2000 and Beyond. Cell. 100(1):113-127. http://dx.doi.org/10.1016/s0092-8674(00)81688-8
2.
Guenther JF, Chanmanivone N, Galetovic MP, Wallace IS, Cobb JA, Roberts DM. 2003. Phosphorylation of Soybean Nodulin 26 on Serine 262 Enhances Water Permeability and Is Regulated Developmentally and by Osmotic Signals. Plant Cell. 15(4):981-991. http://dx.doi.org/10.1105/tpc.009787
2.5 Spacers
Spacers are used to create a distance between the peptide and the cargo to reduce steric hindrance at the binding sites of the peptide. In this case cargo can be a drug, dye, tag.
2.5.1 PEGylation
Attachment of poly (ethylene glycol) to a peptide is called PEGylation. Short bifunctional PEG (Poly (ethylene glycol)) can be used as a spacer in bioconjugation of peptides with other molecules. PEG bioconjugation has also been used to improve proteolytic stability, biodistribution and solubility of peptides1-2.
PEGylation
References
1.
Sullivan TP, van Poll ML, Dankers PYW, Huck WTS. 2004. Forced Peptide Synthesis in Nanoscale Confinement under Elastomeric Stamps. Angew. Chem. Int. Ed.. 43(32):4190-4193. http://dx.doi.org/10.1002/anie.200460271
2.
Veronese FM. 2001. Peptide and protein PEGylation. Biomaterials. 22(5):405-417. http://dx.doi.org/10.1016/s0142-9612(00)00193-9
2.5.2 Amino hexanoic acid
Amino hexanoic acid is a hydrophobic spacer to which a molecule- either a fluorophore, tag or any biological molecule - can be attached to a peptide 1-2.
Amino hexanoic acid
References
1.
Mitchell D, Steinman L, Kim D, Fathman C, Rothbard J. 2000. Polyarginine enters cells more efficiently than other polycationic homopolymers. 56(5):318-325. http://dx.doi.org/10.1034/j.1399-3011.2000.00723.x
2.
Weiler J. 1997. Hybridisation based DNA screening on peptide nucleic acid (PNA) oligomer arrays. 25(14):2792-2799. http://dx.doi.org/10.1093/nar/25.14.2792
3.0 C-Terminal Modifications
3.1 Amide (Amidation)
The C-terminal of the peptide is synthesized as an amide to neutralize negative charge created by the C-terminal COOH. This modification is added to prevent enzyme degradation, to mimic native proteins, and in some cases to remove hydrogen bonding at the C-terminal of the peptides which may interfere with the assays.1
References
1.
Kim K, Seong BL. 2001. Peptide amidation: Production of peptide hormonesin vivo andin vitro. Biotechnol. Bioprocess Eng.. 6(4):244-251. http://dx.doi.org/10.1007/bf02931985