GF65733677

Nickel

foil, 10mm disks, thickness 0.05mm, annealed, 99.98%

Sinónimos:
Nickel, NI000391
Empirical Formula (Hill Notation):
Ni
Número de CAS:
Peso molecular:
58.69
MDL number:
PubChem Substance ID:
NACRES:
NA.23

assay

99.98%

form

foil

manufacturer/tradename

Goodfellow 657-336-77

resistivity

6.97 μΩ-cm, 20°C

diam. × thickness

10 mm × 0.05 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Exclamation markHealth hazard

signalword

Danger

WGK Germany

WGK 1

Flash Point F

Not applicable

Flash Point C

Not applicable

F E Cecil et al.
The Review of scientific instruments, 81(10), 10D326-10D326 (2010-11-09)
We have examined the observed currents in the front foils of the JET Faraday cup lost alpha particle diagnostic KA-2. In particular, we have sought to understand the currents during Ohmic plasmas for which the ion flux at the detectors...
W K Winnett et al.
Talanta, 41(10), 1627-1630 (1994-10-01)
A modification of the sample introduction system for combustion total organic carbon analyzers is presented where sample is sprayed into the instrument. Thermally created sprays and nebulizer sprays are discussed. It is shown that spray introduction gives more symmetric signals...
G E van Dorssen et al.
Journal of X-ray science and technology, 3(2), 109-117 (1992-01-01)
The structure of Ni-C multilayer and single nickel layer samples has been analyzed before and after annealing, using two techniques: fluorescence EXAFS (F1EXAFS) at the Ni-K. edge and CuKα reflection. Annealing at a temperature of 450°C resulted in a change...
R J Schulz et al.
Physics in medicine and biology, 35(11), 1563-1574 (1990-11-01)
The thermal defects of A-150 plastic and graphite referenced to aluminium were determined for 800 keV protons scattered by a 2 micron nickel foil (mean transmitted energy = 550 keV). Composite cores of Al/A-150, graphite/A-150 and Al/graphite which could be...
H T Tien et al.
Photochemistry and photobiology, 49(5), 527-530 (1989-05-01)
Visible light of the solar spectrum is directly converted to stored chemical energy of hydrogen from artificial sea water in a novel electrochemical photovoltaic cell. The principal element of the cell, modeled after the photosynthetic thylakoid membrane, is a semiconductor...

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico

Redes sociales

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Investigación. Desarrollo. Producción.

Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos

© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.

Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.