Merck
All Photos(2)

GF75479057

Nickel

foil, not light tested, 98x100mm, thickness 0.005mm, as rolled, 99.9%

Synonym(s):
Nickel, NI000224
Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
Número MDL:
ID de la sustancia en PubChem:

ensayo

99.9%

formulario

foil

manufacturer/tradename

Goodfellow 754-790-57

resistividad

6.97 μΩ-cm, 20°C

tamaño × grosor

98 x 100 mm × 0.005 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

densidad

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

Related Categories

Descripción general

For updated SDS information please visit www.goodfellow.com.

Información legal

Product of Goodfellow

pictogramas

Exclamation markHealth hazard

Palabra de señalización

Danger

Frases de peligro

Clasificaciones de peligro

Carc. 2 - Skin Sens. 1 - STOT RE 1

Código de clase de almacenamiento

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service