WHA2814533
Whatman® high-purity glass microfiber extraction thimbles
HP, L × W 53 mm × 135 mm
Sinónimos:
Z753556
packaging
pk of 25
manufacturer/tradename
Whatman 2814-533
Whatman Article No. 28419030 (US reference)
L × W
53 mm × 135 mm
¿Está buscando productos similares? Visit Guía de comparación de productos
General description
Whatman® High-Purity Glass Microfiber Extraction Thimbles
High-purity glass microfiber thimbles are available for specialized applications. The thimbles are completely free of binders or additives and can be used at temperatures up to 500°C or when using solvents incompatible with cellulose thimbles. These thimbles are also used in pollution monitoring techniques (0.8 μm nominal particle retention). Typical thickness 1.7 mm.
Features and benefits:
  • Available in a range of sizes and wall thicknesses to suit your application
  • Designed to fit most commercially available Soxhlet extractors
  • No binders are added
Applications:
  • Smoke stack gas monitoring
  • Soxhlet extraction
  • Analyzing pesticide residues
  • Determining oil/fat content of foods (e.g. French fries)
  • Analysis of oil and grease in solid wastes
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva