Z236128
Supelco
Ferrule Remover
NACRES:
SB.54
availability
available only in EU
Categorías relacionadas
General description
Spiral tapered tip for removal of capillary ferrules from nuts. Two sizes are included, for removing 0.4 mm and 0.8 mm I.D. ferrules.
Certificado de Análisis
Certificado de origen