Z236136
Supelco
Septum Pick
NACRES:
SB.54
Categorías relacionadas
General description
The septum pick is useful for removing small pieces of septa from injection ports, and graphite ferrules from column nuts.
Certificado de Análisis
Certificado de origen