Merck
All Photos(1)

12607

Supelco

Chlortoluron-(N,N-dimethyl-d6)

PESTANAL®, analytical standard

Synonym(s):
3-(3-Chloro-4-methyl)-1,1-dimethyl-d6-urea, Chlortoluron-d6
Empirical Formula (Hill Notation):
C10D6H7ClN2O
Molecular Weight:
218.71
Número MDL:
ID de la sustancia en PubChem:

grado

analytical standard

Nivel de calidad

pureza isotópica

≥99 atom % D

Línea del producto

PESTANAL®

ensayo

≥98.0% (HPLC)

caducidad

limited shelf life, expiry date on the label

application(s)

agriculture

formato

neat

SMILES string

CC1=C(Cl)C=C(NC(N(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)C=C1

InChI

1S/C10H13ClN2O/c1-7-4-5-8(6-9(7)11)12-10(14)13(2)3/h4-6H,1-3H3,(H,12,14)/i2D3,3D3

InChI key

JXCGFZXSOMJFOA-XERRXZQWSA-N

Related Categories

Descripción general

Chlortoluron-(N,N-dimethyl-d6) is an isotope labeled analog of the phenylurea herbicide chlortoluron, wherein the N,N-dimethyl protons are replaced by deuterium.

Aplicación

Chlortoluron-(N,N-dimethyl-d6) may be used as an internal standard for the determination of antifouling pesticides by high performance liquid chromatography (HPLC) coupled to tandem mass spectrometry (MS/MS).
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis, potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Envase

5 mg

Información legal

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

pictogramas

Health hazardEnvironment

Palabra de señalización

Warning

Frases de peligro

Clasificaciones de peligro

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Código de clase de almacenamiento

11 - Combustible Solids

WGK

WGK 3

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service