Merck
All Photos(1)

81475

Supelco

Poly(butyl methacrylate)

analytical standard, for GPC, 800,000

CAS Number:
Número MDL:
ID de la sustancia en PubChem:
NACRES:
NA.24

grado

analytical standard
for GPC

Nivel de calidad

caducidad

limited shelf life, expiry date on the label

mol peso

Mn ~682000
Mp ~772000
Mw ~723000

technique(s)

gel permeation chromatography (GPC): suitable

densidad

1.07 g/mL at 25 °C (lit.)

Mw/Mn

~1.06

formato

neat

SMILES string

CCCCOC(=O)C(C)=C

InChI

1S/C8H14O2/c1-4-5-6-10-8(9)7(2)3/h2,4-6H2,1,3H3

InChI key

SOGAXMICEFXMKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Código de clase de almacenamiento

13 - Non Combustible Solids

WGK

WGK 3

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Equipo de protección personal

Eyeshields, Gloves, type N95 (US)

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service