Merck
All Photos(1)

CL13

CCS Viscosity Standard

ASTM D 5293 and SAE Specification J307

Synonym(s):
Cold-Cranking Simulator Viscosity Standard
NACRES:
NA.24

grado

certified reference material

caducidad

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

formato

mixture

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

1 of 4

This Item
CL48CL09CL60
vibrant-m

CL13

CCS Viscosity Standard

vibrant-m

CL48

CCS Viscosity Standard

vibrant-m

CL09

CCS Viscosity Standard

vibrant-m

CL60

CCS Viscosity Standard

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

format

mixture

format

mixture

format

mixture

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

Descripción general

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon CCS Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Código de clase de almacenamiento

10 - Combustible liquids

WGK

WGK 1

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service