Merck
All Photos(1)

PE1038

Supelco

Trace Metals (Dissolved) PT - 1

Proficiency Testing Material

NACRES:
NA.24

Nivel de calidad

envase

ampule of 2 × 20 mL

concentración

in nitric acid

application(s)

environmental

formato

matrix material

Compare Similar Items

View Full Comparison

1 of 4

This Item
NIST3140NIST1643FSQC005
vibrant-m

NIST3140

Platinum (Pt) standard solution

vibrant-m

NIST1643F

Trace Elements in Water

Supelco

Supelco

SQC005

TCLP Metals in Soil

application(s)

environmental

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

environmental

format

matrix material

format

single component solution

format

-

format

matrix material

packaging

ampule of 2 × 20 mL

packaging

pkg of 5 x 10 mL

packaging

pkg of 250 mL

packaging

-

concentration

in nitric acid

concentration

-

concentration

-

concentration

-

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

300

Descripción general

Trace Metals (Dissolved) PT - 1 proficiency testing material is produced in accordance with ISO/IEC 17043:2010. This product is also referred to as non-Potable water or Water Pollution sample. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Most water PT samples are available as concentrates or ready-to-use volumes giving the end-user flexibility in selecting samples that will fit their exact needs. Our WP program is designed to meet the quality assurance requirements, thereby ensuring the analytical competence of your laboratory.
These samples are typically provided in duplicate for use in case of lab mishap, for QC purposes or to analyze multiple methods.

Type of PT Scheme: Water analysis - Water Pollution (WP)

Matrix:
Water Pollution/Wastewater

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of Trace Metals (Dissolved) PT - 1 can be found in the reporting packet which will be available to download at PT Portal.

Aplicación

Trace Metals (Dissolved) PT - 1 is a non-Potable water (NPW) PT standard, suitable for testing dissolved trace metals in water pollution/wastewater.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Características y beneficios

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Nota de preparación

Description

  • This PT sample is packaged as a 20 mL concentrate.
  • Definition of a dissolved metal: Those metals in an unacidified sample that pass through a 0.45 μm membrane filter.
  • The solvent for this sample is 1% nitric acid by volume in DI water. The sample is slightly acidified to maintain stability in the concentrate and will not interfere with the method. Please follow preparation directions for best performance.
  • The PT sample has not been preserved.

Preparation Instructions

  • To prepare the solution, partially fill a 1000 mL volumetric flask with reagent-grade water (near 20 ºC).
  • Shake ampule well prior to opening.
  • Open ampule PE1038-20ML (be careful to avoid cuts on glass) and transfer 10.0 mL of concentrate to the volumetric flask.
  • Fill the flask to volume with reagent-grade water and mix well.
  • The sample is now ready for immediate analysis. Please note that the sample has not been acidified, you will need to acidify as required by your method after filtration.
  • A reagent-grade blank should be analyzed concurrently for background correction. The water blank should be made to match the acid matrix of the standards and samples.

Almacenamiento y estabilidad

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Otras notas

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2022-2023)

Wastewater
WP22-2 March23 - May 6, 2022
WP22-4 September 21 - November 4, 2022

WP23-2 March22 – May 5, 2023
WP23-4 September 20 – November 3, 2023


Quick-Turn Study (on demand proficiency test that is offereddepending on lot availability)

Información legal

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Aluminum, Al 200-4000 μg/L
    Arsenic, As 70-900 μg/L
    Beryllium, Be 8-900 μg/L
    Cadmium, Cd 8-2500 μg/L
    Chromium, Cr (total) 17-3000 μg/L
    Cobalt, Co 28-2500 μg/L
    Copper, Cu 40-3000 μg/L
    Iron, Fe 200-4000 μg/L
    Lead, Pb 70-4000 μg/L
    Lithium, Li 0-4000 μg/L
    Manganese, Mn 70-4000 μg/L
    Mercury, Hg 2-30 μg/L
    Nickel, Ni 80-3500 μg/L
    Selenium, Se 90-2000 μg/L
    Vanadium, V 55-6000 μg/L
    Zinc, Zn 100-2000 μg/L
    See All (16)

Palabra de señalización

Danger

Clasificaciones de peligro

Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Código de clase de almacenamiento

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service