Merck
All Photos(1)

PE1205

Supelco

Mercury (Low Level) - WP

Proficiency Testing Material

Synonym(s):
WP - Mercury (Low Level)
NACRES:
NA.24

descripción

Concentrate for dilution to 1L

Nivel de calidad

envase

ampule of 2 × 20 mL

concentración

in nitric acid

application(s)

environmental

formato

matrix material

Descripción general

Mercury (Low Level) - WP proficiency testing material is produced in accordance with ISO/IEC 17043:2010. Also referred to as non-Potable water or Water Pollution sample, this sample is produced according to TNI requirements. However, please note, this sample may include non-TNI analytes as well. The higher criteria from the TNI tables mimic the levels common to wastewater samples. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Most water PT samples are available as concentrates or ready-to-use volumes giving the end-user flexibility in selecting samples that will fit their exact needs. Our WP program is designed to meet the quality assurance requirements in compliance with Discharge Monitoring Report - Quality Assurance Program (DMR-QA), thereby ensuring the analytical competence of your laboratory, thereby ensuring the analytical competence of your laboratory.
These samples are typically provided in duplicate for use in case of lab mishap, for QC purposes or to analyze multiple methods.

Type of PT Scheme: Water analysis - Water Pollution (WP)

Matrix: Water Pollution/Wastewater

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of Mercury (Low Level) - WP can be found in the reporting packet which will be available to download at PT Portal.

Aplicación

Mercury (Low Level) - WP is a non-Potable water (NPW) PT standard, suitable for mercury testing in water pollution/wastewater.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Características y beneficios

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Nota de preparación

Description

  • This PT sample is packaged as a 20 mL concentrate.
  • The solvent for this sample is 5% nitric acid by volume in DI water. This also acts as the preservative.

Preparation Instructions

  • Partially fill a 1000 mL volumetric flask with reagent-grade water (near 20 ºC) and add 1 mL of trace-metal-grade nitric acid.
  • Open ampule PE1205-20ML (be careful to avoid cuts on glass) and transfer 10 mL of concentrate to the volumetric flask.
  • Fill the flask to volume with reagent-grade water and mix well.
  • The sample is now ready for immediate analysis.
  • A reagent-grade blank should be analyzed concurrently for background correction. the water blank should be made to match the acid matrix of the standards and samples.

NOTE: The 0.15% v/v nitric acid specification is a minimum; the amount and type of acid may be altered to routine analytical procedures if needed.
NOTE: Mercury is present as inorganic mercury.

Almacenamiento y estabilidad

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Otras notas

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2022-2023)


Wastewater
WP22-2 March 23 - May 6, 2022

WP23-2 March 22 – May 5, 2023

DMR-QA (US customers only)
DMR-QA 43 2023 Dates TBD

Quick-Turn Study (on demand proficiency test that is offereddepending on lot availability)

Información legal

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Low Level Mercury 20-100 ng/L

pictogramas

Corrosion

Palabra de señalización

Danger

Frases de peligro

Clasificaciones de peligro

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Código de clase de almacenamiento

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service