UBBEL14UKC
Ubbelohde Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
Size 4C, Nom. Constant 30.0, Range 6,000 to 30,000 cSt
NACRES:
NA.24
manufacturer/tradename
Paragon Scientific Ltd
Featured Industry
Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care
¿Está buscando productos similares? Visit Guía de comparación de productos
General description
This product is issued with a UKAS 17025 certificate of calibration