Merck
All Photos(2)

SAB4501386

Sigma-Aldrich

Anti-SOX8 antibody produced in rabbit

affinity isolated antibody

Synonym(s):
Transcription factor SOX-8
Número MDL:
NACRES:
NA.41

origen biológico

rabbit

conjugado

unconjugated

forma del anticuerpo

affinity isolated antibody

antibody product type

primary antibodies

clon

polyclonal

formulario

buffered aqueous solution

mol peso

antigen 47 kDa

species reactivity

mouse, human

concentración

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

Nº de acceso NCBI

Nº de acceso UniProt

enviado en

wet ice

temp. de almacenamiento

−20°C

Gene Information

human ... SOX8(30812)

Descripción general

Anti-SOX8 Antibody detects endogenous levels of total SOX8 protein.

Inmunógeno

The antiserum was produced against synthesized peptide derived from human SOX8/9/17/18.

Immunogen Range: 116-165

Características y beneficios

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Forma física

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Cláusula de descargo de responsabilidad

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Código de clase de almacenamiento

12 - Non Combustible Liquids

WGK

nwg

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service