Merck
All Photos(1)

SBP4933

Sigma-Aldrich

JPH4 (AB2) Blocking Peptide for PRS4933

NACRES:
NA.41

Nivel de calidad

100

origen biológico

synthetic

formulario

liquid

species reactivity

human, rat, mouse

concentración

200 μg/mL

enviado en

wet ice

temp. de almacenamiento

−20°C

Inmunógeno

17 amino acids near the center of human JPH4.

Aplicación

Used as a blocking peptide in immunoblotting applications.

Ligadura / enlace

This peptide blocks the action of antibody PRS4933.

Cláusula de descargo de responsabilidad

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Código de clase de almacenamiento

12 - Non Combustible Liquids

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service