Merck
All Photos(1)

SRP4142

Sigma-Aldrich

IL-5 from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):
BCDFII, Eosinophil differentiation factor, TRF, B cell differentiation factor I, T-cell replacing factor, IL-5
Número MDL:
NACRES:
NA.32

origen biológico

rat

recombinante

expressed in E. coli

ensayo

≥98% (HPLC)
≥98% (SDS-PAGE)

formulario

lyophilized

mol peso

~13.1 kDa

envase

pkg of 10 μg

impurezas

endotoxin, tested

Nº de acceso NCBI

enviado en

wet ice

temp. de almacenamiento

−20°C

Gene Information

rat ... Il5(24497)

Descripción general

IL-5, also known as eosinophil differentiation factor. It regulates eosinophil growth and activation. The recombinant IL-5 is a dimeric, non-glycosylated polypeptide with 113 amino acids.

Forma física

Lyophilized from 1 mg/ml in sterile solution with no additives.

Reconstitución

Reconstitute in water to a concentration of 0.1-0.5 mg/mL. The solution can be diluted into other aqueous buffers.
Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service