SRP4991

Sigma-Aldrich

Prolactin Receptor from rat

recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)

Sinónimos:
PRL-R, Prlr, Prolactin receptor, Lactogen receptor
En este momento no podemos mostrarle ni los precios ni la disponibilidad

biological source

rat

recombinant

expressed in E. coli

assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

mol wt

~24.12 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Prlr(24684)

General description

Prolactin is a pituitary hormone that plays a role in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The primary step in its action is the binding to a specific membrane receptor (prolactin receptor) which belongs to the superfamily of class 1 cytokine receptors. Prolactin is a hormone involved in a range of significant functions including ion transport and osmoregulation, stimulation of milk, protein synthesis as well as the regulation of numerous reproductive functions. Prolactin exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane Prolactin receptor. PRLR varies in size (short and long forms) with tissue source and species, from ~40 kDa to 100 kDa. Recombinant human Prolactin Receptor (Extra Cellular Domain) produced in E. coli is a non-glycosylated, Polypeptide chain containing 206 amino acids and having a molecular mass of 24.12 kDa.

Physical form

Lyophilized from a concentrated (1 mg/ml) solution with 0.0045 mM NaHCO3.

Reconstitution

Reconstitute in sterile dH2O to a concentration of 0.1 -1 mg/mL and let the lyophilized pellet dissolve completely. This solution can then be diluted into other aqueous buffers.
Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point F

Not applicable

Flash Point C

Not applicable

Certificado de Análisis

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico

Redes sociales

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Investigación. Desarrollo. Producción.

Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos

© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.

Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.