SRP5019

Sigma-Aldrich

DCAMKL1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sinónimos:
KIAA0369, DCDC3A, DCLK, DCLK1
NACRES:
NA.32
En este momento no podemos mostrarle ni los precios ni la disponibilidad

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

2.9-3.9 nmol/min·mg

mol wt

~110 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DCLK1(9201)

General description

DCAMKL1 or doublecortin-like kinase 1 contains two N-terminal doublecortin domains (which bind microtubules and regulate microtubule polymerization), a C-terminal serine/threonine protein kinase domain (which shows substantial homology to Ca2+/calmodulin-dependent protein kinase), and a serine/proline-rich domain in between the doublecortin and the protein kinase domains (which mediates multiple protein-protein interactions). DCAMKL1 is a microtubule-associated kinase that can undergo autophosphorylation. DCAMKL1 has microtubule-polymerizing activity that is independent of its protein kinase activity. DCAMKL1 is involved in several different cellular processes, including neuronal migration, retrograde transport, neuronal apoptosis and neurogenesis.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point F

Not applicable

Flash Point C

Not applicable

Shogo Ohmae et al.
The Journal of biological chemistry, 281(29), 20427-20439 (2006-05-11)
Despite the critical importance of Ca(2+)/calmodulin (CaM)-dependent protein kinase (CaMK) II signaling in neuroplasticity, only a limited amount of work has so far been available regarding the presence and significance of another predominant CaMK subfamily, the CaMKI/CaMKIV family, in the...
P T Lin et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 20(24), 9152-9161 (2000-01-11)
Doublecortin (DCX) is a microtubule-associated protein required for neuronal migration to the cerebral cortex. DCAMKL1 consists of an N terminus that is 65% similar to DCX throughout the entire length of DCX, but also contains an additional 360 amino acid...

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico

Redes sociales

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Investigación. Desarrollo. Producción.

Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos

© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.

Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.