Merck
All Photos(1)
SRP5201
Sigma-Aldrich
KAT9, GST tagged human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
FLJ10422
NACRES:
NA.32
recombinante
expressed in baculovirus infected Sf9 cells
ensayo
≥70% (SDS-PAGE)
formulario
buffered aqueous glycerol solution
mol peso
~86 kDa
Nº de acceso NCBI
enviado en
dry ice
temp. de almacenamiento
−70°C
Gene Information
human ... ELP3(55140)
Descripción general
KAT9 (also known as ELP3) is the catalytic subunit of the histone acetyltransferase (HAT) elongator complex, which contributes to transcript elongation and also regulates the maturation of projection neurons. The knockdown of KAT9 by antisense morpholinos in zebrafish embryos resulted in dose-dependent shortening and abnormal branching of motor neurons with no concomitant morphologic abnormalities. KAT9 knockdown also impairs paternal DNA demethylation as indicated by reporter binding, immunostaining, and bisulfite sequencing.
Forma física
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Nota de preparación
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Código de clase de almacenamiento
10 - Combustible liquids
WGK
WGK 1
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Quotes and Ordering
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service