Merck
All Photos(1)

SRP6189

Sigma-Aldrich

GRO/KC murine

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):
Gro1, KC, N51, CXCL1, Growth-regulated α protein, Scyb1, Secretory protein N51, chemokine (C-X-C motif) ligand 1, Mgsa, gro, Fsp, Platelet-derived growth factor-inducible protein KC

origen biológico

mouse

recombinante

expressed in E. coli

ensayo

≥98% (SDS-PAGE)

formulario

lyophilized

mol peso

7.8 kDa

envase

pkg of 100 μg

Nº de acceso UniProt

enviado en

wet ice

temp. de almacenamiento

−20°C

Gene Information

mouse ... GROA(14825)

Descripción general

Murine KC (also known as murine GRO-α) belongs to the C-X-C family of chemokines. Themurine KC is a 7.8 kDa protein containing 72 amino acid residues.

Forma física

Sterile filtered and lyophilized with no additives.

Reconstitución

Reconstitute in H₂O to a concentration of 0.1-1 mg/mL. This solution can be diluted into other aqueous buffers or stored at 4°C for 1 week or –20°C for future use.

Código de clase de almacenamiento

13 - Non Combustible Solids

WGK

WGK 3

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service