Merck
All Photos(1)

SRP6221

Sigma-Aldrich

IFN-γ1 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):
IL-29
NACRES:
NA.32

origen biológico

human

recombinante

expressed in E. coli

ensayo

≥98% (SDS-PAGE)

formulario

lyophilized powder

mol peso

19.8 kDa

envase

pkg of 10 and 50 μg

impurezas

<1 EU/μg endotoxin (LAL test)

Nº de acceso UniProt

enviado en

wet ice

temp. de almacenamiento

−20°C

Gene Information

human ... IFNL1(282618)

Descripción general

ICAMs are members of the Ig superfamily of calcium-independent transmembrane glycoproteins. ICAM-1 is a ligand for lymphocyte function-associated (LFA) and Mac-1 integrins and the major human rhinovirus receptor. The primary function of ICAM-1 is to provide adhesion between endothelial cells and leukocytes after stress or injury. The human ICAM-1 gene codes for a 505 amino acid transmembrane glycoprotein containing a 29 amino acid cytoplasmic domain, a 23 amino acid transmembrane domain, and a 453 amino acid extracellular domain. Recombinant human ICAM-1 is a 49.5 kDa glycoprotein comprising the extracellular domain (453 amino acid residues) of ICAM-1. Monomeric glycosylated ICAM-1 migrates at an apparent molecular weight of approximately 72.0-80.0 kDa by SDS-PAGE analysis under reducing conditions.

Forma física

Sterile filtered through a 0.2 micron filter. Lyophilized from 10 mM Sodium Phosphate, pH 7.5.

Reconstitución

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.5-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Código de clase de almacenamiento

10 - Combustible liquids

WGK

WGK 1

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service