Merck
All Photos(1)

SRP6356

Sigma-Aldrich

CD83 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):
BL11, CD83 antigen isoform b precursor, HB15
NACRES:
NA.32

origen biológico

human

recombinante

expressed in E. coli

ensayo

≥90% (SDS-PAGE)

formulario

liquid

mol peso

16 kDa (148 aa, 20-144 aa + His Tag)

envase

pkg of 50 μg

concentración

0.25 mg/mL

Nº de acceso UniProt

enviado en

dry ice

temp. de almacenamiento

−70°C

Gene Information

human ... CD83(9308)

Descripción general

CD83 is a single-pass type I membrane protein and member of the immunoglobulin superfamily of receptors. The protein may be involved in the regulation of antigen presentation. A soluble form of this protein can bind to dendritic cells and inhibit their maturation. Three transcript variants encoding different isoforms have been found for this gene. Recombinant human CD83 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.

Forma física

0.25 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.15 M NaCl and 30% glycerol.

Código de clase de almacenamiento

13 - Non Combustible Solids

WGK

WGK 3

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service