SRP6471

Sigma-Aldrich

HVEM/TNFRSF14 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Sinónimos:
HVEA, Tumor necrosis factor receptor superfamily member 14, Herpesvirus entry mediator., TR2, ATAR, LIGHTR
NACRES:
NA.32
En este momento no podemos mostrarle ni los precios ni la disponibilidad

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

monomer observed mol wt 50-60 kDa by SDS-PAGE (reducing) (DTT-reduced.)
calculated mol wt 45.4 kDa (This protein is a disulfide-linked homodimeric protein after removal of the signal peptide. The 413 aa reduced monomer is fused at the C-terminus to human IgG1 (rhHVEM-Fc).)

packaging

pkg of 20 and 100 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFRSF14(8764)

General description

Herpesvirus entry mediator (HVEM), also known as TNFRSF14, TR2 (TNF receptor like molecule) and ATAR (another TRAF associated receptor), is a type I membrane protein belonging to the TNF/NGF receptor superfamily. HVEM expression has been detected in peripheral blood T cells, B cells, monocytes and in various tissues enriched in lymphoid cells. The extracellular domain of HVEM has been shown to interact directly with the herpes simplex virus envelope glycoprotein D (gD). Two TNF superfamily ligands, including the secreted TNFβ (lymphotoxin α) and the membrane protein LIGHT (lymphotoxins, exhibits inducible expression, and competes with HSV glycoprotein D for HVEM, a receptor expressed by T lymphocytes), have been shown to be the cellular ligands for HVEM. Besides HVEM, LIGHT can also interact with LTβR, the receptor for lymphotoxin αβ heterotrimer. The role of the HVEM LIGHT /LTβ receptor ligand pair in immune function and herpesvirus pathobiology remains to be elucidated.

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM tris, 100 mM glycine, pH 7.0. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point F

Not applicable

Flash Point C

Not applicable

Certificado de Análisis

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico

Redes sociales

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Investigación. Desarrollo. Producción.

Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos

© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.

Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.