Merck
All Photos(1)

SRP6536

Sigma-Aldrich

RBP4 from rat

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):
PRBP, Plasma retinol-binding protein, RBP, Retinol Binding Protein 4
NACRES:
NA.32

origen biológico

rat

recombinante

expressed in E. coli

ensayo

≥90% (SDS-PAGE)

formulario

liquid

mol peso

monomer ~230 kDa (Rat RBP4 aa 19-201 is fused at the C-terminus to a His-Tag.)

envase

pkg of 50 μg

impurezas

<0.1 EU/μg endotoxin (LAL test)

Nº de acceso UniProt

enviado en

wet ice

temp. de almacenamiento

−20°C

Gene Information

rat ... RBP4(25703)

Descripción general

Retinol binding protein 4 (RBP4; RBP) is a 21 kDa secreted protein, a member of the lipocalin family and is known as the primary transporter of retinol (vitamin A) to tissues. A recent report revealed RBP4 as an adipokine linking glucose transporter 4 (GLUT4) suppression in adipose tissue to insulin. Elevated human and mouse serum RBP4 levels are associated with insulin resistance and its severity, obesity, and certain components of metabolic syndrome. Furthermore, human serum RBP4 levels are closely related to renal function.

Forma física

0.2 μm-filtered solution in 55 mM TRIS-Cl, pH 8.2 containing 150 mM NaCl. .

Código de clase de almacenamiento

10 - Combustible liquids

WGK

WGK 1

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service