22447
Supelco
Packed Column Filling Kit
Categorías relacionadas
General description
This convenient kit contains many of the items need to quickly and efficiently pack columns. Includes two puller/inserter tools (Cat. No. 22406), plastic funnel, rubber column connector, and 50 g of silanized glass wool for plugs.
Certificado de Análisis
Certificado de origen