23640-U
Supelco
Conversion Kit for Adapting Packed GC to Capillary GC
port diam. 5 mm, sleeve L 5 in. (13 cm)
NACRES:
SB.54
port diam.
5 mm
sleeve L
5 in. (13 cm)
compatibility
for connecting Direct Injection Conversion Kit
¿Está buscando productos similares? Visit Guía de comparación de productos
General description
These column nuts are replacements for damaged or misplaced original Agilent/HP MSD nuts. They are to be used with the longer design ferrules for Agilent/HP GC′s.
With our Make-up Gas Detector Adapter Kit you can use the capillary column at optimum flow rates (2-10mL/minute), even if your detector is designed for higher flows. You can connect the column and a make-up gas line (to prevent column effluent from spreading in the detector) to the detector without modifying your detector gas line. Regulate the make-up gas through external control (see diagram) or, in a two-column instrument, connect the make-up gas line to the second injection port and regulate flow with the flow controller (requires a Swagelok reducer � Cat. No. 22017-U for a 1/8 in. fitting, Cat. No. 22701 for a 1/4 in. fitting � and nut; order separately).
Application
Conversion kit maybe used to quantitatively determine ethanol concentration in alcoholic beverages using capillary gas chromatography.
Other Notes
Use with long Supeltex ferrules.
Certificado de Análisis
A rapid method for determination of ethanol in alcoholic beverages using capillary gas chromatography.
Wang ML
Journal of Food Science, 11 (2), 133-140 (2003)
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.