Merck
All Photos(1)
2631205
Supelco
Inlet Liner, Direct (SPME) Type, Straight Design (unpacked)
technique used for SPME, pkg of 5 ea
packaging
pkg of 5 ea
L × O.D. × I.D.
92 mm × 6.35 mm × 0.75 mm
compatibility
configured for PerkinElmer Auto System, Model 9000 Split/Splitless Capillary Injector
technique used for SPME
Compare Similar Items
View Full Comparison
1 of 4
This Item
263642526339012637501
packaging
pkg of 5 ea
packaging
pkg of 25 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
L × O.D. × I.D.
92 mm × 6.35 mm × 0.75 mm
L × O.D. × I.D.
54 mm × 4.6 mm × 0.8 mm
L × O.D. × I.D.
95 mm × 5.0 mm × 0.75 mm
L × O.D. × I.D.
78.5 mm × 6.5 mm × 0.75 mm
compatibility
configured for PerkinElmer Auto System, Model 9000 Split/Splitless Capillary Injector
compatibility
-
compatibility
for use with Shimadzu 17A with SPL-17 Injectors, technique used for SPME
compatibility
-
General description
Inlet liner is an important part within the injection port. The prime functions of inlet liner are to limit sample degradation and improve vaporisation along with correct positioning of syringe needle. A wrong choice of liner can have prominent effect on the chromatogram. The basic problems related with incorrect choice of liner include the higher potential of peak tailing and mass discrimination. The inlet liner volume is recommended to be larger than the amount of sample injected by syringe so as to avoid any reaction with the sample. Injector port liner is suitable to make connection between sample introduction and GC column. Direct type has narrow bore which sustains a high linear velocity through the injection port to diminish band broadening and transferring all the samples to column, flash vaporisation.
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits PerkinElmer (AutoSystem)
Application
Inlet liner, direct type may be used for solid phase microextraction (SPME) sample injection.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
(enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service