47652-U

Supelco

EPA 552.1 Acids Calibration Mix with Surrogate

certified reference material, in methyl tert-butyl ether (varied)

EC Number:
NACRES:
NA.24

Quality Level

grade

certified reference material
TraceCERT®

Agency/Method

EPA 552

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

in methyl tert-butyl ether (varied)

application(s)

HPLC: suitable
gas chromatography (GC): suitable

Featured Industry

Environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analito

Descripción

    Bromoacetic acid 200 μg/mL
    Bromochloroacetic acid 200 μg/mL
    2-Bromopropionic acid 100 μg/mL
    Chloroacetic acid 300 μg/mL
    Dibromoacetic acid 100 μg/mL
    Dichloroacetic acid 300 μg/mL
    2,2-Dichloropropionic acid 200 μg/mL
    Trichloroacetic acid 100 μg/mL
    Ver todo (8)

Certificado de Análisis

Certificado de origen

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico