Merck
All Photos(1)

1358809

USP

Levalbuterol Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):
4-[2-(tert-Butylamino)-1-hydroxyethyl]-2-methylphenol, α-{[(1,1-Dimethylethyl)amino]methyl}-4-hydroxy-3-methylbenzenemethanol
Empirical Formula (Hill Notation):
C13H21NO2
CAS Number:
Molecular Weight:
223.31
NACRES:
NA.24

grado

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

formato

neat

Descripción general

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Nota de análisis

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Otras notas

USP issued SDS can be found here.
Sales restrictions may apply.

pictogramas

Exclamation mark

Palabra de señalización

Warning

Frases de peligro

Clasificaciones de peligro

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1B

Código de clase de almacenamiento

11 - Combustible Solids

WGK

WGK 3

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service