Merck
All Photos(1)

1715000

USP

Viomycin sulfate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):
Viomycin sulfate salt, Tuberactinomycin B sulfate salt
Empirical Formula (Hill Notation):
C25H43N13O10 · H2SO4
CAS Number:
Molecular Weight:
783.77
Número MDL:
ID de la sustancia en PubChem:

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

SMILES string

[O-]S([O-])(=O)=O.NCCC[C@H]([NH3+])CC(=O)N[C@H]1CNC(=O)[C@@H](NC(=O)\C(NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC1=O)=C\NC(N)=O)[C@H]2C[C@H](O)NC([NH3+])=N2

InChI

1S/C25H43N13O10.H2O4S/c26-3-1-2-10(27)4-16(41)32-12-6-30-23(47)18(11-5-17(42)37-24(28)36-11)38-20(44)13(7-31-25(29)48)33-21(45)14(8-39)35-22(46)15(9-40)34-19(12)43;1-5(2,3)4/h7,10-12,14-15,17-18,39-40,42H,1-6,8-9,26-27H2,(H,30,47)(H,32,41)(H,33,45)(H,34,43)(H,35,46)(H,38,44)(H3,28,36,37)(H3,29,31,48);(H2,1,2,3,4)/b13-7-;/t10-,11+,12-,14-,15-,17-,18-;/m0./s1

InChI key

AQONYROJHRNYQQ-QMAPKBLTSA-N

Looking for similar products? Visit Product Comparison Guide

Descripción general

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Nota de análisis

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Otras notas

USP issued SDS can be found here.
Sales restrictions may apply.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service