Nombre del producto
PDGF-BB human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Referencia del producto
SRP3138
Marca de producto
SIGMA
Número de CAS
Peso molecular
Ensayo
Especificación
AppearanceReport result
ColorReport result
Purity by SDS-Page≥98%
Purity by HPLC≥98%
Endotoxin Level≤1 EU/µg
Biological ActivityDetermined by its ability to stimulate the proliferation of mouse Balb/c 3T3 cells. The expected ED50 for this effect is 1.0-3.0 ng/mL.