Nombre del producto
Granzyme B from mouse recombinant, expressed in Hi-5 Insect cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture
Referencia del producto
SRP3202
Marca de producto
SIGMA
Número de CAS
Peso molecular
Ensayo
Especificación
AppearanceReport result
ColorWhite to off-white
Purity by SDS-Page≥97%
Purity by HPLC≥97%
Endotoxin Level≤ 1 EU/µg
Biological ActivityDetermined by its ability to cleave a synthetic chromogentic Granzyme B substrate. The expected specific activity, when using the Ac-IEPD-pNA substrate at 25 °C, is greater than 750 nM/min per mg of enzyme.