Nombre del producto
FGF-4 human Animal-component free, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
Referencia del producto
SRP3261
Marca de producto
SIGMA
Número de CAS
Peso molecular
Ensayo
Especificación
AppearanceReport result
colorReport result
Purity by SDSPage≥95%
Purity by HPLC≥95%
Endotoxin Level≤1 EU/µg
Biological ActivityDetermined by the dosedependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors. The expected ED50 is ≥ 0.5 ng/mL, corresponding to a specific activity of > 2 x 106 units/mg