Nombre del producto
IFN-LAMBDA1 human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Referencia del producto
SRP3273
Marca de producto
SIGMA
Número de CAS
Peso molecular
Ensayo
Especificación
Sourcerecombinant, expressed in E. coli
Molecular Weight19.8 kDa
Storage-20°C
ReconstitutionCentrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Appearancelyophilized powder
Colorwhite to off-white
Purity by HPLC>=98%
Purity by SDS-PAGE>=98%
Endotoxin Level<= 1 EU/ug
Biological ActivityDetermined by its ability to activate STAT phosphorylation in an ISRE Luciferase Reporter assay using human colon carcinoma (COLO205) cells. The expected ED50 is 0.2-0.5 ng/ml.