Nombre del producto
NGF-beta from mouse recombinant, expressed in E. coli, ≥98% (SDS-PAGE)
Referencia del producto
SRP4304
Marca de producto
SIGMA
Número de CAS
Peso molecular
Ensayo
Especificación
AppearanceLyophilized protein
SDS-PAGE>=98%
HPLCNot Verified
Endotoxin<0.1 ng per mg by LAL method
Biological ActivityActivity was measured in a cell proliferation assay using a factor-dependent human erythroleukemic cells line. The ED50 for this effect is typically 0.2 ng/ml