Nombre del producto
Clusterin (nuclear form) mouse recombinant, expressed in E. coli, His tagged, >90% (SDS-PAGE)
Referencia del producto
SRP8003
Marca de producto
SIGMA
Número de CAS
Peso molecular
Ensayo
Especificación
Sourcerecombinant, expressed in E. coli
Molecular Weight~45 kDa (SDS-PAGE)
Storage/Stability-20�C. Working aliquots are stable for up to 3 months when stored at -20�C. Avoid repeated freeze/thaw cycles.
FormulationSolution in 55 mM TRIS-HCl, pH 8.2, containing 150 mM NaCl.
Formliquid
Colorclear solution
Concentration>=0.2 mg/ml
Purity by SDS-PAGE>=90%
Endotoxin Level<1 EU/ug purified protein (LAL test).