Nombre del producto
Lipase from Candida lipolytica powder (fine), ≥5 U/g
Referencia del producto
62303
Marca de producto
sial
Número de CAS
9001-62-1
Peso molecular
Ensayo
Especificación
APPEARANCE (COLOR)White to Beige
APPEARANCE (FORM)Fibers
SOLUBILITY (COLOR)for information
SOLUBILITY (TURBIDITY)for information
SOLUBILITY (METHOD)5 MG/ML IN H2O
ENZYMATIC ACTIVITY≥ 5.0 U/g
UNIT DEFINITION1 U CORRESPONDS TO THE AMOUNT OF ENZYME WHICH HYDROLYSES 1 UMOL of p-NITROPHENYL BUTYRATE PER MINUTE AT 25 C IN 50 mM TRIS-HCl BUFFER, at pH7.5, 10% ACETONITRILE.
RECOMMENDED RETEST PERIOD48 MONTHS