MX
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
Amino-functionalized silica gel spherical
(1)
Amino-functionalized silica gel spherical
CAS Number:
126850-14-4
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
79297
Amino-functionalized silica gel spherical, 40-75 μm particle size
Expand
View Pricing