MX
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
Carbonyl cyanide 3-chlorophenylhydrazone
(1)
Carbonyl cyanide 3-chlorophenylhydrazone
Synonyms:
m
-Cl-CCP, CCCP, Mesoxalonitrile 3-chlorophenylhydrazone
Empirical Formula (Hill Notation):
C
9
H
5
ClN
4
CAS Number:
555-60-2
Molecular Weight:
204.62
EC Number:
209-103-7
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
C2759
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Expand
View Pricing