MX
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
ISA (ionic strength adjustment solution: 1 M KCl)
(1)
ISA (ionic strength adjustment solution: 1 M KCl)
Synonyms:
Potassium chloride solution
Linear Formula:
KCl
CAS Number:
7447-40-7
Molecular Weight:
74.55
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
58221
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Expand
View Pricing