MX
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
L-Xylulose 5-phosphate lithium salt
(1)
L-Xylulose 5-phosphate lithium salt
Empirical Formula (Hill Notation):
C
5
H
11
O
8
P · xLi
+
CAS Number:
18050-82-3
Molecular Weight:
230.11 (free acid basis)
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
00993
L-Xylulose 5-phosphate lithium salt,
BioChemika
, ≥90% (TLC)
Expand
View Pricing