Myosin1D is an evolutionarily conserved regulator of animal left-right asymmetry.
Nature communications (2018-05-18)
Thomas Juan, Charles Géminard, Jean-Baptiste Coutelis, Delphine Cerezo, Sophie Polès, Stéphane Noselli, Maximilian Fürthauer
RESUMEN
The establishment of left-right (LR) asymmetry is fundamental to animal development, but the identification of a unifying mechanism establishing laterality across different phyla has remained elusive. A cilia-driven, directional fluid flow is important for symmetry breaking in numerous vertebrates, including zebrafish. Alternatively, LR asymmetry can be established independently of cilia, notably through the intrinsic chirality of the acto-myosin cytoskeleton. Here, we show that Myosin1D (Myo1D), a previously identified regulator of Drosophila LR asymmetry, is essential for the formation and function of the zebrafish LR organizer (LRO), Kupffer's vesicle (KV). Myo1D controls the orientation of LRO cilia and interacts functionally with the planar cell polarity (PCP) pathway component VanGogh-like2 (Vangl2), to shape a productive LRO flow. Our findings identify Myo1D as an evolutionarily conserved regulator of animal LR asymmetry, and show that functional interactions between Myo1D and PCP are central to the establishment of animal LR asymmetry.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-Acetylated Tubulin antibody produced in mouse, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Ouabain, European Pharmacopoeia (EP) Reference Standard
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.