A Short BRCA2-Derived Cell-Penetrating Peptide Targets RAD51 Function and Confers Hypersensitivity toward PARP Inhibition.
Molecular cancer therapeutics (2018-04-15)
Anika Trenner, Julia Godau, Alessandro A Sartori
RESUMEN
Under conditions of genotoxic stress, cancer cells strongly rely on efficient DNA repair to survive and proliferate. The human BRCA2 tumor suppressor protein is indispensable for the repair of DNA double-strand breaks by homologous recombination (HR) by virtue of its ability to promote RAD51 loading onto single-stranded DNA. Therefore, blocking the interaction between BRCA2 and RAD51 could significantly improve the efficacy of conventional anticancer therapies. However, targeting protein-protein interaction (PPI) interfaces has proven challenging because flat and large PPI surfaces generally do not support binding of small-molecule inhibitors. In contrast, peptides are more potent for targeting PPIs but are otherwise difficult to deliver into cells. Here, we report that a synthetic 16-mer peptide derived from the BRC4 repeat motif of BRCA2 is capable of blocking RAD51 binding to BRCA2. Efficient noncytotoxic cellular uptake of a nona-arginine (R9)-conjugated version of the BRC4 peptide interferes with DNA damage-induced RAD51 foci formation and HR. Moreover, transduction of the BRC4 peptide impairs replication fork-protective function of BRCA2 and triggers MRE11-dependent degradation of nascent DNA in response to DNA replication stress. Finally, the BRC4 cell-penetrating peptide (CPP) confers selective hypersensitivity to PARP inhibition in cancer cells but spares noncancerous cells. Taken together, our data highlight an innovative approach to develop novel peptide-based DNA repair inhibitors and establish BRCA2-derived CPPs as promising anticancer agents. Mol Cancer Ther; 17(7); 1392-404. ©2018 AACR.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95%
Sigma-Aldrich
RAD51 Inhibitor B02, ≥98% (HPLC)
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.