MX
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
Structure of the alpha beta tubulin dimer by electron crystallography.
Structure of the alpha beta tubulin dimer by electron crystallography.
Nature (1998)
Nogales E