Merck
MK-2206, an allosteric inhibitor of AKT, stimulates LDLR expression and LDL uptake: A potential hypocholesterolemic agent.
Atherosclerosis (2018-07-20)
Katrine Bjune, Hilde Sundvold, Trond P Leren, Soheil Naderi
RESUMEN
Induction of low-density lipoprotein receptor (LDLR) plays a significant role in reduction of plasma LDL-cholesterol (LDL-C) levels. Therefore, strategies that enhance the protein level of LDLR provide an attractive therapeutic target for the treatment of hypercholesterolemia. With this aim in mind, we concentrated our effort on studying the role of AKT kinase in regulation of LDLR levels and proceeded to examine the effect of MK-2206, an allosteric and highly selective AKT inhibitor, on LDLR expression. Cultured human hepatoma cells were used to examine the effect of MK-2206 on the proteolytic processing of sterol regulatory element-binding protein-2 (SREBP-2), the expression of LDLR and cellular internalization of LDL. We also examined the effect of MK-2206 on LDLR levels in primary human hepatocytes. MK-2206 induced the proteolytic processing of SREBP-2, upregulated LDLR expression and stimulated LDL uptake. In contrast to statins, induction of LDLR levels by MK-2206 did not rely on 3-hydroxy-3-methylglutaryl-CoA reductase (HMGCR) inhibition. As a result, cotreatment of cells with MK-2206 and mevastatin potentiated the impact of mevastatin on LDLR. Importantly, MK-2206 stimulated the expression of LDLR by primary human hepatocytes. MK-2206 is a novel LDLR-inducing agent that, either alone or in combination with statins, exerts a stimulating effect on cellular LDL uptake.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%
Sigma-Aldrich
Monoclonal Anti-GAPDH−Peroxidase antibody produced in mouse, clone GAPDH-71.1, purified immunoglobulin, buffered aqueous solution